1-Bromo-2-butyne ≥99%

427292-5G 427292-1G
220066-483EA 149.69 USD
220066-483 220066-482
1-Bromo-2-butyne ≥99%
1-Bromo-2-butyne
Formula: C₄H₅Br
MW: 132.99 g/mol
MDL Number: MFCD00190233
CAS Number: 3355-28-0
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR