Dimethylamine 40% (w/w) in aqueous solution

Danger

426458-2L 426458-1L 426458-100ML
220066-451EA 83.08 USD
220066-451 220066-450 220066-449
Dimethylamine 40% (w/w) in aqueous solution
Dimethylamine
Formula: C₂H₇N
MW: 45.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008288
CAS Number: 124-40-3
EINECS: 204-697-4
UN: 1160
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR