Tetrabutylammonium hydroxide 1 M in aqueous solution ACS

426326-100ML 426326-25ML
220066-440EA 175.38 USD
220066-440 220066-441
Tetrabutylammonium hydroxide 1 M in aqueous solution ACS
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00009425
CAS Number: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR