Tetramethylammonium bromide ACS

Danger

426296-25G 426296-100G
220066-437EA 36.54 USD
220066-437 220066-436
Tetramethylammonium bromide ACS
Tetramethylammonium bromide
Formula: C₄H₁₂BrN
MW: 154.05 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR