Tetrabutylammonium bromide ACS

426288-100G 426288-25G
220066-369EA 70.62 USD
220066-369 220066-435
Tetrabutylammonium bromide ACS
Tetrabutylammonium bromide
Formula: C₁₆H₃₆BrN
MW: 322.37 g/mol
Melting Pt: 100…103 °C
Density: 1.148 g/cm³ (20 °C)
MDL Number: MFCD00011633
CAS Number: 1643-19-2
EINECS: 216-699-2

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR