2-Butynoic acid ethyl ester ≥98%

Warning

Synonyms: Ethyl 2-butynoate

425117-1G 425117-10G
220066-300EA 40.38 USD
220066-300 220066-299
2-Butynoic acid ethyl ester ≥98%
2-Butynoic acid ethyl ester
Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00015182
CAS Number: 4341-76-8
EINECS: 224-395-6
UN: 3272
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR