3-Aminoisoxazole

424218-5ML 424218-25ML
220066-256EA 179.85 USD
220066-256 220066-255
3-Aminoisoxazole
3-Aminoisoxazole
Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD00038814
CAS Number: 1750-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR