1-Butyl-3-methylimidazolium thiocyanate, Sigma-Aldrich®

42254-100G-F 42254-1KG-F
220066-136EA 856 USD
220066-136 220066-137
1-Butyl-3-methylimidazolium thiocyanate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium thiocyanate
Formula: C₉H₁₅N₃S
MW: 197.3 g/mol
MDL Number: MFCD06798181
CAS Number: 344790-87-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR