(1S,2S)-(+)-trans-1,2-Cyclohexanediol ≥99%

Synonyms: (1S,2S)-(+)-trans-Cyclohexane-1,2-diol, (1S,2S)-trans-1,2-Cyclohexanediol

421804-250MG 421804-1G
220066-080EA 126 USD
220066-080 220066-079
(1S,2S)-(+)-trans-1,2-Cyclohexanediol ≥99%
(1S,2S)-(+)-trans-1,2-Cyclohexanediol
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 108 °C
CAS Number: 57794-08-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR