Talc

Warning

420433-500G 420433-2.5KG
220065-917EA 473.38 USD
220065-917 220065-916
Talc
Talc
Formula: Mg₃O₁₁Si₄·1H₂O
Melting Pt: ? 1200 °C
Density: 2.58 to 2.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00792903
CAS Number: 14807-96-6
EINECS: 238-877-9
Merck Index: 13,09127

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR