5-Methylanthranil acid ≥99%

Warning

Synonyms: 2-Amino-5-methylbenzoic acid, 6-Amino-m-toluic acid

419443-25G 419443-5G
220065-847EA 464.15 USD
220065-847 220065-848
5-Methylanthranil acid ≥99%
5-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 317 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR