Pentadecan-15-olide

Synonyms: Cyclopentadecanolide, Oxacyclohexadecan-2-one, Pentadecanolide, ω-Pentadecalactone, omega-Pentadecalactone, Oxa-2-cyclohexadecanone

419133-25G 419133-100G
220065-829EA 71.08 USD
220065-829 220065-828
Pentadecan-15-olide
Pentadecan-15-olide
Formula: C₁₅H₂₈O₂
MW: 240.39 g/mol
MDL Number: MFCD00039667
CAS Number: 106-02-5
EINECS: 203-354-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR