4-Methoxyphenylboronic acid

Warning

Synonyms: 4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid

417599-5G 417599-1G 417599-25G
220065-600EA 54.28 USD
220065-600 220065-598 220065-599
4-Methoxyphenylboronic acid
4-Methoxyphenylboronic acid
Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR