4-Fluorophenylboronic acid

Warning

Synonyms: (4-Fluorophenyl)metaboric acid, 4-Fluorobenzeneboronic acid, Dihydroxy(4-fluorophenyl)borane

417556-25G 417556-1G 417556-5G
220065-594EA 74.77 USD
220065-594 220065-593 220065-595
4-Fluorophenylboronic acid
4-Fluorophenylboronic acid
Formula: C₆H₆BFO₂
MW: 139.92 g/mol
Melting Pt: 263…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00039136
CAS Number: 1765-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR