4-Chlorophenylboronic acid ≥95%

Warning

Synonyms: 4-Chlorobenzeneboronic acid, Dihydroxy(4-chlorophenyl)borane, (4-Chlorophenyl)boronic acid, (4-Chlorophenyl)metaboric acid

417548-10G 417548-25G 417548-1G
220065-590EA 16.62 USD
220065-590 220065-592 220065-591
4-Chlorophenylboronic acid ≥95%
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 268…272 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR