3-Chlorophenylboronic acid

Warning

Synonyms: 3-Chlorobenzeneboronic acid, (3-Chlorophenyl)metaboric acid, 3-Chlorophenyl boronic acid, Dihydroxy(3-chlorophenyl)borane

417521-10G 417521-1G
220065-588EA 134.31 USD
220065-588 220065-589
3-Chlorophenylboronic acid
3-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00161354
CAS Number: 63503-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR