5-Bromo-2,4-dichloropyrimidine

416762-25G 416762-5G
220065-540EA 145.38 USD
220065-540 220065-541
5-Bromo-2,4-dichloropyrimidine
5-Bromo-2,4-dichloropyrimidine
Formula: C₄HBrCl₂N₂
MW: 227.87 g/mol
Boiling Pt: 288 °C (1013 hPa)
Melting Pt: -1 °C
Density: 1.781 g/cm³ (20 °C)
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR