Dimethyl sulfide

Danger

Synonyms: DMS

41625-250ML 41625-1L
220065-508EA 71.86 USD
220065-508 220065-507
Dimethyl sulfide
Dimethyl sulphide
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 36.2 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3,II
Merck Index: 12,06204

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR