1-Ethylpiperazine

Synonyms: N-Ethylpiperazine

415308-250ML 415308-50ML
220065-424EA 149.69 USD
220065-424 220065-425
1-Ethylpiperazine
1-Ethylpiperazine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.8989 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00059912
CAS Number: 5308-25-8
EINECS: 226-166-6
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR