Strontium hydroxide octahydrate ≥95%

Danger

415219-1KG 415219-500G
220064-274EA 124.62 USD
220064-274 220064-275
Strontium hydroxide octahydrate ≥95%
Strontium hydroxide octahydrate
Formula: H₂O₂Sr·8H₂O
MW: 265.76 g/mol
Melting Pt: 375 °C
Density: 1.9 g/cm³ (20 °C)
MDL Number: MFCD00150395
CAS Number: 1311-10-0
EINECS: 242-367-1
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR