Strontium oxide ≥99.9%

415138-50G 415138-10G
220064-014EA 381.54 USD
220064-014 220064-013
Strontium oxide ≥99.9%
Strontium oxide
Formula: SrO
MW: 103.62 g/mol
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 2430 °C
Density: 4.7 g/cm³ (20 °C)
MDL Number: MFCD00049558
CAS Number: 1314-11-0
EINECS: 215-219-9
UN: 3262
ADR: 8 II
Merck Index: 12,09009

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR