Magnesium stearate, Technical Grade

415057-25G 415057-1KG
220065-414EA 47.71 USD
220065-414 220063-685
Magnesium stearate, Technical Grade
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MDL Number: MFCD00036391
CAS Number: 557-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR