Hexyl isocyanate ≥97%

Synonyms: Isocyanic acid hexyl ester

414883-5ML 414883-25ML
220065-410EA 120.62 USD
220065-410 220065-409
Hexyl isocyanate ≥97%
Hexyl isocyanate
Formula: C₇H₁₃NO
MW: 127.19 g/mol
Boiling Pt: 44 °C (1013 hPa)
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00037047
CAS Number: 2525-62-4
EINECS: 219-763-8
UN: 3080
ADR: 6.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR