Barium iodide, anhydrous ≥99.995% (trace metals basis), beads −10 mesh

Warning

413615-25G 413615-5G
220065-331EA 1106.15 USD
220065-331 220065-332
Barium iodide, anhydrous ≥99.995% (trace metals basis), beads −10 mesh
Barium iodide
Formula: BaI₂
MW: 391.14 g/mol
Melting Pt: 714 °C
Density: 5.15 g/cm³ (20 °C)
MDL Number: MFCD00003452
CAS Number: 13718-50-8
EINECS: 237-276-9
UN: 1564
ADR: 6.1,III
Merck Index: 12,01009

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR