2-Fluorobenzoic acid ≥97%

Warning

Synonyms: o-Fluorobenzoic acid

412244-100G 412244-25G
220065-206EA 86.58 USD
220065-206 220065-207
2-Fluorobenzoic acid ≥97%
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 121 - 124 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR