3-Bromo-p-anisaldehyde ≥98%

Synonyms: 3-Bromo-4-methoxybenzaldehyde

412015-25G 412015-5G
220065-195EA 204.92 USD
220065-195 220065-196
3-Bromo-p-anisaldehyde ≥98%
3-Bromo-p-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 108…110 °C (1 mmHg)
Melting Pt: 54 °C
MDL Number: MFCD00016599
CAS Number: 34841-06-0
EINECS: 252-241-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR