N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99.5%

Danger

Synonyms: 1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine

411019-1L 411019-100ML
220065-132EA 468.46 USD
220065-132 220065-131
N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99.5%
N,N,N',N'-Tetramethylethylenediamine (TEMED)
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: -64 °C
Density: 0.7765 g/cm³ (20 °C)
Flash Pt: 17 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
EINECS: 203-744-6
UN: 2372
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR