α-Bromo-3-(trifluoromethoxy)toluene ≥97%

Danger

408891-5G 408891-1G
220064-950EA 206.31 USD
220064-950 220064-949
α-Bromo-3-(trifluoromethoxy)toluene ≥97%
α-Bromo-3-(trifluoromethoxy)toluene
Formula: C₈H₆BrF₃O
Boiling Pt: 178 - 180 °C (1013 hPa)
Density: 1.572 g/cm³ (20 °C)
MDL Number: MFCD00061271
CAS Number: 159689-88-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR