1-Benzyl-3-methylimidazolium tetrafluoroborate

Synonyms: 1-Benzyl-3-methyl-1H-imidazol-3-ium tetrafluoroborate

40819-5G-F 40819-50G-F
220064-909EA 120.77 USD
220064-909 220064-027
1-Benzyl-3-methylimidazolium tetrafluoroborate
1-Benzyl-3-methylimidazolium tetrafluoroborate
Formula: C₁₁H₁₃BF₄N₂
MW: 260.04 g/mol
MDL Number: MFCD07784449
CAS Number: 500996-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR