3-Methoxy-3-methyl-1-butanol ≥98%

Synonyms: 3-Methyl-3-methoxybutanol, Solvant 39, MMB, 3-Methoxy-3-methylbutan-1-ol

408069-250ML 408069-1L
220064-908EA 61.28 USD
220064-908 220064-907
3-Methoxy-3-methyl-1-butanol ≥98%
3-Methoxy-3-methyl-1-butanol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.9038 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR