1,3-Dimethyl-2-imidazolidinone

Synonyms: 1,3-Dimethylimidazolidin-2-one

40727-100ML 40727-1L
220064-782EA 34.26 USD
220064-782 220064-783
1,3-Dimethyl-2-imidazolidinone
1,3-Dimethyl-2-imidazolidinone
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 225.5 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.04 g/cm³ (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00003188
CAS Number: 80-73-9
EINECS: 201-304-8
UN: 2810
ADR: 6.1,III
Merck Index: 13,03275

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR