Diethyl pyrocarbonate ≥99%

Warning

Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC

40718-100ML 40718-25ML
220064-773EA 570.77 USD
220064-773 220064-774
Diethyl pyrocarbonate ≥99%
Diethyl pyrocarbonate
Formula: O(COOC₂H₅)₂
MW: 162.14 g/mol
Boiling Pt: 93 °C (1013 hPa)
Density: 1.123 g/cm³ (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009106
CAS Number: 1609-47-8
EINECS: 216-542-8
Merck Index: 13,08088

Order Now

Specification Test Results

Assay 99% NT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR