2-Amino-4-chlorobenzonitrile ≥99%

Warning

Synonyms: 5-Chloro-2-cyanoaniline

405280-10G 405280-50G
220064-596EA 81.74 USD
220064-596 220064-597
2-Amino-4-chlorobenzonitrile ≥99%
2-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00035927
CAS Number: 38487-86-4
EINECS: 253-967-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR