Titanium(IV) methoxide

Synonyms: Titanium tetramethanolate, Titanium (IV) methoxide

404950-10G 404950-50G
220064-586EA 87.85 USD
220064-586 220064-587
Titanium(IV) methoxide
Titanium(IV) methoxide
Formula: C₄H₁₂O₄Ti
MW: 172.02 g/mol
MDL Number: MFCD00014883
CAS Number: 992-92-7
EINECS: 213-596-4
UN: 3181
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR