1-Fluoro-4-ethynylbenzene ≥99%

404330-5G 404330-1G
220064-534EA 97.25 USD
220064-534 220064-533
1-Fluoro-4-ethynylbenzene ≥99%
1-Fluoro-4-ethynylbenzene
Formula: C₈H₅F
Flash Pt: <61 °C
MDL Number: MFCD00168823
CAS Number: 766-98-3
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR