1,3-Dimethylurea, Technical Grade

Synonyms: N,N'-Dimethylurea

40410-1KG 40410-50G
220064-511EA 49.54 USD
220064-511 220064-512
1,3-Dimethylurea, Technical Grade
1,3-Dimethylurea
Formula: C₃H₈N₂O
MW: 88.11 g/mol
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 96 °C
Density: 1.142 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008286
CAS Number: 96-31-1
EINECS: 202-498-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR