Strontium ≥99%

403326-25G 403326-5G
220064-462EA 404.62 USD
220064-462 220064-463
Strontium ≥99%
Strontium
Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1151 °C (1013 hPa)
Melting Pt: 570 - 660 °C
Density: 2.38 g/cm³ (770 °C)
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3,II
Merck Index: 12,08994

Order Now

Specification Test Results

Assay 99% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR