Sodium chlorate ≥99.0% ACS

Danger

403016-100G 403016-500G
220064-424EA 52.12 USD
220064-424 220064-425
Sodium chlorate ≥99.0% ACS
Sodium chlorate
Formula: ClNaO₃
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 260 °C
Density: 2.498 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
EINECS: 231-887-4
UN: 1495
ADR: 5.1,II
REACH: 01-2119474389-23
Merck Index: 13,08670

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR