Lead(II) oxide ≥99.0% ACS

Danger

402982-1KG 402982-250G
220064-414EA 159.38 USD
220064-414 220064-415
Lead(II) oxide ≥99.0% ACS
Lead(II) oxide
Formula: PbO
Boiling Pt: 1470 °C (1013 hPa)
Melting Pt: 888 °C
Density: 9.5 g/cm³ (20 °C)
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1,III
Merck Index: 13,05431

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR