p-Toluenesulfonic acid monohydrate ≥98.5% ACS

Warning

Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate

402885-5G 402885-100G 402885-500G
220064-385EA 24.62 USD
220064-385 220064-383 220064-384
p-Toluenesulfonic acid monohydrate ≥98.5% ACS
p-Toluenesulphonic acid monohydrate
Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,III
Merck Index: 12,09671

Order Now

Specification Test Results

Assay 98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR