Calcium acetate monohydrate ≥99.0% ACS

402850-100G 402850-500G
220064-377EA 43.45 USD
220064-377 220064-378
Calcium acetate monohydrate ≥99.0% ACS
Calcium acetate monohydrate
Formula: (H₃CCOO)₂Ca·1H₂O
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150019
CAS Number: 5743-26-0
EINECS: 200-540-9
Merck Index: 13,01645

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR