Acetaldehyde ≥99.5% ACS

Danger

Synonyms: Ethanal

402788-5ML 402788-1L 402788-100ML
220063-966EA 28.46 USD
220063-966 220063-965 220063-964
Acetaldehyde ≥99.5% ACS
Acetaldehyde
Formula: CH₃CHO
MW: 44.05 g/mol
Boiling Pt: 20.1 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.78 g/cm³ (20 °C)
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006991
CAS Number: 75-07-0
EINECS: 200-836-8
UN: 1089
ADR: 3,I
Merck Index: 12,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR