4-(Diphenylphosphino)benzoic acid ≥97%

401595-1G 401595-5G
220064-247EA 63 USD
220064-247 220064-248
4-(Diphenylphosphino)benzoic acid ≥97%
4-(Diphenylphosphino)benzoic acid
Formula: C₁₉H₁₅O₂P
MW: 306.3 g/mol
MDL Number: MFCD00407264
CAS Number: 2129-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR