Potassium hydrogen sulfate ≥99.99%

Danger

Synonyms: Potassium bisulphate

401544-50G 401544-10G
220064-240EA 293.85 USD
220064-240 220064-239
Potassium hydrogen sulfate ≥99.99%
Potassium hydrogen sulphate
Formula: KHSO₄
MW: 136.17 g/mol
Melting Pt: 214 °C
Density: 2.32 g/cm³ (20 °C)
MDL Number: MFCD00011404
CAS Number: 7646-93-7
EINECS: 231-594-1
UN: 2509
ADR: 8,II
Merck Index: 13,07695

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR