Zinc perchlorate hexahydrate

401439-100G 401439-500G
220064-228EA 58.62 USD
220064-228 220064-229
Zinc perchlorate hexahydrate
Zinc perchlorate hexahydrate
Formula: Cl₂O₈Zn·6H₂O
Melting Pt: 106 °C
Density: 2.25 g/cm³ (20 °C)
MDL Number: MFCD00150370
CAS Number: 10025-64-6
EINECS: 237-122-0
UN: 1481
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR