cis-2-Butene ≥99%

Danger

400890-25G 400890-100G
220061-977EA 232.62 USD
220061-977 220061-976
cis-2-Butene ≥99%
cis-2-Butene
Formula: C₄H₈
MW: 56.11 g/mol
Boiling Pt: 3.7 °C (1013 hPa)
Melting Pt: -139.3 °C
Density: 0.6213 g/cm³ (20 °C)
CAS Number: 590-18-1
EINECS: 209-673-7
UN: 1012
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR