Tantalum pentachloride

400475-25G 400475-5G
220064-194EA 280 USD
220064-194 220064-195
Tantalum pentachloride
Tantalum pentachloride
Formula: Cl₅Ta
Boiling Pt: 232…235 °C (1013 hPa)
Melting Pt: 216…217 °C
Density: 3.68 g/cm³ (20 °C)
MDL Number: MFCD00011253
CAS Number: 7721-01-9
EINECS: 231-755-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR