Cobalt(II) acetate ≥99.995%

Danger

Synonyms: Cobaltous acetate, Cobalt acetate, Acetic acid cobalt (II) salt, Cobalt diacetate

399973-10G 399973-1G
220064-181EA 296.92 USD
220064-181 220064-182
Cobalt(II) acetate ≥99.995%
Cobalt(II) acetate
Formula: C₄H₆CoO₄
MW: 177.02 g/mol
Melting Pt: 297 - 299 °C
Density: 1.7755 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008689
CAS Number: 71-48-7
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 12,02494

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR