1-Butyl-3-methylimidazolium acetate, Sigma-Aldrich®

39952-1KG-F 39952-100G-F
220064-155EA 2120 USD
220064-155 220064-154
1-Butyl-3-methylimidazolium acetate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium acetate
Formula: C₁₀H₁₈N₂O₂
MDL Number: MFCD06798175
CAS Number: 284049-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR