1-Methylbenzimidazole

Synonyms: 1-Methyl-1H-benzimidazole

399353-25G 399353-5G
220064-144EA 137.85 USD
220064-144 220064-145
1-Methylbenzimidazole
1-Methylbenzimidazole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 59…62 °C
Density: 1.11 g/cm³ (25 °C)
MDL Number: MFCD00192275
CAS Number: 1632-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR